Merck
  • Microbiome as Mediator of Diet on Colorectal Cancer Risk: The Role of Vitamin D, Markers of Inflammation and Adipokines.
Microbiome as Mediator of Diet on Colorectal Cancer Risk: The Role of Vitamin D, Markers of Inflammation and Adipokines.
Nutrients (2021-01-29)
Davide Serrano, Chiara Pozzi, Silvia Guglietta, Bruno Fosso, Mariano Suppa, Patrizia Gnagnarella, Federica Corso, Federica Bellerba, Debora Macis, Valentina Aristarco, Paolo Manghi, Nicola Segata, Cristina Trovato, Maria Giulia Zampino, Marinella Marzano, Bernardo Bonanni, Maria Rescigno, Sara Gandini
ABSTRACT
Obesity and diet are associated with colorectal cancer (CRC) risk, and microbiome could mediate this risk factor. To investigate this interaction, we performed a case-control study (34 CRC cases and 32 controls) and analyzed fecal microbiota composition using 16S rRNA metabarcoding and sub-sequential shotgun analyses of genomic bacterial DNA to evaluate the role of microbiome and diet in CRC etiology, taking into account vitamin D and other risk biomarkers. Dietary habits were evaluated using a short questionnaire. Multivariate methods for data integration and mediation analysis models were used to investigate causal relationships. CRC cases were significantly more often deficient in vitamin D than controls (p = 0.04); FokI and CYP24A1 polymorphism frequency were different between cases and controls (p = 0.03 and p = 0.02, respectively). A diet poor in fatty fish and rich in carbohydrates was found to be significantly associated with CRC risk (p = 0.011). The mediation analysis confirmed the significant role of the microbiome in mediating CRC risk-increasing levels of Bifidobacteria/Escherichia genera ratio, an indicator of "healthy" intestinal microbiome, can overcome the effect of diet on CRC risk (p = 0.03). This study suggests that microbiome mediates the diet effect on CRC risk, and that vitamin D, markers of inflammation, and adipokines are other factors to consider in order to achieve a better knowledge of the whole carcinogenic process.
Sigma-Aldrich
Mouse IGF-II ELISA Kit, for cell culture supernatant
Pricing and availability is not currently available.