Merck
  • Molecular Mechanism of HER2 Rapid Internalization and Redirected Trafficking Induced by Anti-HER2 Biparatopic Antibody.
Molecular Mechanism of HER2 Rapid Internalization and Redirected Trafficking Induced by Anti-HER2 Biparatopic Antibody.
Antibodies (Basel, Switzerland) (2020-09-24)
Jackie Cheng, Meina Liang, Miguel F Carvalho, Natalie Tigue, Raffaella Faggioni, Lorin K Roskos, Inna Vainshtein
ABSTRACT
Amplification and overexpression of HER2 (human epidermal growth factor receptor 2), an ErbB2 receptor tyrosine kinase, have been implicated in human cancer and metastasis. A bispecific tetravalent anti-HER2 antibody (anti-HER2-Bs), targeting two non-overlapping epitopes on HER2 in domain IV (trastuzumab) and domain II (39S), has been reported to induce rapid internalization and efficient degradation of HER2 receptors. In this study, we investigated the molecular mechanism of this antibody-induced rapid HER2 internalization and intracellular trafficking. Using quantitative fluorescent imaging, we compared the internalization kinetics of anti-HER2-Bs and its parental arm antibodies, alone or in combinations and under various internalization-promoting conditions. The results demonstrated that concurrent engagement of both epitopes was necessary for rapid anti-HER2-Bs internalization. Cellular uptake of anti-HER2-Bs and parental arm antibodies occurred via clathrin-dependent endocytosis; however, inside the cells antibodies directed different trafficking pathways. Trastuzumab dissociated from HER2 in 2 h, enabling the receptor to recycle, whereas anti-HER2-Bs stayed associated with the receptor throughout the entire endocytic pathway, promoting receptor ubiquitination, trafficking to the lysosomes, and efficient degradation. Consistent with routing HER2 to degradation, anti-HER2-Bs significantly reduced HER2 shedding and altered its exosomal export. Collectively, these results enable a better understanding of the mechanism of action of anti-Her2-Bs and can guide the rational design of anti-HER2 therapeutics as well as other bispecific molecules.
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
MG-132, InSolution, ≥98%, 10 mM, reversible proteasome inhibitor