Merck
A termination-independent role of Rat1 in cotranscriptional splicing.
Nucleic acids research (2021-05-13)
Zuzer Dhoondia, Hesham Elewa, Marva Malik, Zahidur Arif, Roger Pique-Regi, Athar Ansari
ABSTRACT
Rat1 is a 5'→3' exoribonuclease in budding yeast. It is a highly conserved protein with homologs being present in fission yeast, flies, worms, mice and humans. Rat1 and its human homolog Xrn2 have been implicated in multiple nuclear processes. Here we report a novel role of Rat1 in mRNA splicing. We observed an increase in the level of unspliced transcripts in mutants of Rat1. Accumulation of unspliced transcripts was not due to the surveillance role of Rat1 in degrading unspliced mRNA, or an indirect effect of Rat1 function in termination of transcription or on the level of splicing factors in the cell, or due to an increased elongation rate in Rat1 mutants. ChIP-Seq analysis revealed Rat1 crosslinking to the introns of a subset of yeast genes. Mass spectrometry and coimmunoprecipitation revealed an interaction of Rat1 with the Clf1, Isy1, Yju2, Prp43 and Sub2 splicing factors. Furthermore, recruitment of splicing factors on the intron was compromised in the Rat1 mutant. Based on these findings we propose that Rat1 has a novel role in splicing of mRNA in budding yeast. Rat1, however, is not a general splicing factor as it crosslinks to only long introns with an average length of 400 nucleotides.
Sigma-Aldrich
Anti-RNA Polymerase II Antibody, CTD Antibody, clone 8WG16, clone 8WG16, from mouse