Merck
  • The influence of sleep apnea syndrome and intermittent hypoxia in carotid adventitial vasa vasorum.
The influence of sleep apnea syndrome and intermittent hypoxia in carotid adventitial vasa vasorum.
PloS one (2019-02-06)
Carolina López-Cano, Ferran Rius, Enric Sánchez, Anna Michela Gaeta, Àngels Betriu, Elvira Fernández, Andree Yeramian, Marta Hernández, Marta Bueno, Liliana Gutiérrez-Carrasquilla, Mireia Dalmases, Albert Lecube
ABSTRACT
Subjects with sleep apnea-hypopnea syndrome (SAHS) show an increased carotid intima-media thickness. However, no data exist about earlier markers of atheromatous disease, such as the proliferation and expansion of the adventitial vasa vasorum (VV) to the avascular intima in this setting. Our aim was to assess carotid VV density and its relationship with sleep parameters in a cohort of obese patients without prior vascular events. A total of 55 subjects evaluated for bariatric surgery were prospectively recruited. A non-attended respiratory polygraphy was performed. The apnea-hypopnea index (AHI) and the cumulative percentage of time spent with oxygen saturation below 90% (CT90) were assessed. Serum concentrations of soluble intercellular adhesion molecule 1, P-selectin, lipocalin-2 and soluble vascular cell adhesion molecule 1 (sVCAM-1) were measured. Contrast-enhanced carotid ultrasound was used to assess the VV density. Patients with SAHS (80%) showed a higher adventitial VV density (0.801±0.125 vs. 0.697±0.082, p = 0.005) and higher levels of sVCAM-1 (745.2±137.8 vs. 643.3±122.7 ng/ml, p = 0.035) than subjects with an AHI lower than 10 events/hour. In addition, a positive association exist between mean VV density and AHI (r = 0.445, p = 0.001) and CT90 (r = 0.399, p = 0.005). Finally, in the multiple linear regression analysis, female sex, fasting plasma glucose and AHI (but not CT90) were the only variables independently associated with the mean adventitial VV density (R2 = 0.327). In conclusion, a high VV density is present in obese subjects with SAHS, and chronic intermittent hypoxia is pointed as an independent risk factor for the development of this early step of atheromatous disease.
Product Description
Millipore
MILLIPLEX® Human Cardiovascular Disease Panel, Configurable Human CVD 10-Plex Panel 2