Merck
  • Generation of a human induced pluripotent stem cell line CERAi001-A-6 using episomal vectors.
Generation of a human induced pluripotent stem cell line CERAi001-A-6 using episomal vectors.
Stem cell research (2017-09-28)
Raymond C B Wong, Sandy S Hung, Stacey Jackson, Vikrant Singh, Shahnaz Khan, Helena H Liang, Lisa S Kearns, Tu Nguyen, Alison Conquest, Maciej Daniszewski, Alex W Hewitt, Alice Pébay
ABSTRACT
We report the generation of the hiPSC line CERAi001-A-6 from primary human dermal fibroblasts. Reprogramming was performed using episomal vector delivery of OCT4, SOX2, KLF4, L-MYC, LIN28 and shRNA for p53.
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-AFP Mouse mAb (1G7), liquid, clone 1G7, Calbiochem®
Pricing and availability is not currently available.