Merck
  • Long non-coding RNA PGM5-AS1 promotes epithelial-mesenchymal transition, invasion and metastasis of osteosarcoma cells by impairing miR-140-5p-mediated FBN1 inhibition.
Long non-coding RNA PGM5-AS1 promotes epithelial-mesenchymal transition, invasion and metastasis of osteosarcoma cells by impairing miR-140-5p-mediated FBN1 inhibition.
Molecular oncology (2020-05-16)
Wei Liu, Pengcheng Liu, Hang Gao, Xu Wang, Ming Yan
ABSTRACT
Osteosarcoma is an uncommon tumor occurring in bone, accompanied by elevated incidence and reduced rate of healing. Epithelial-to-mesenchymal transition (EMT) serves as a conceptual paradigm to explain the invasion and metastasis of osteosarcoma and other cancers. Hence, developing effective therapeutic strategy to treat the EMT of osteosarcoma is essential. Here, we identified the molecular mechanism of long noncoding RNA (lncRNA) PGM5-AS1 in EMT and progression of osteosarcoma. Microarray-based analysis was employed to screen the osteosarcoma-related differentially expressed lncRNAs. The levels of PGM5-AS1 as well as microRNA-140-5p (miR-140-5p) and fibrillin-1 (FBN1) in osteosarcoma tissues and cells were determined. Dual-luciferase reporter gene assay, RNA pull-down assay, and RNA immunoprecipitation assay were conducted to validate the relationship among PGM5-AS1, miR-140-5p, and FBN1. Expression of PGM5-AS1, miR-140-5p, and FBN1 was altered by overexpression, shRNA, mimic, or inhibitors in order to investigate how they regulated migration, invasion, and EMT of osteosarcoma cells in vitro. Loss- and gain-of-function approaches were employed in nude mice to detect their roles in tumorigenesis in vivo. Osteosarcoma tissues and cells exhibited low expression of miR-140-5p, but high expression of PGM5-AS1 and FBN1. PGM5-AS1 competitively bound to miR-140-5p to upregulate FBN1. Furthermore, hindering PGM5-AS1 and FBN1 or overexpressing miR-140-5p dampened migration, invasion, and EMT of osteosarcoma cells in vitro. Furthermore, silencing PGM5-AS1 or FBN1, or overexpressing miR-140-5p markedly inhibited tumorigenesis in nude mice in vivo. Taken together, PGM5-AS1 depletion causes FBN1 reduction to retard osteosarcoma processes by negatively modulating miR-140-5p.
Product Description
Roche
SP6/T7 Transcription Kit, sufficient for 2 x 20 assays (stadard transcription), kit of 1 (12 components), suitable for DNA sequencing, suitable for hybridization
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep