Merck
  • CDK4/6 inhibition blocks cancer metastasis through a USP51-ZEB1-dependent deubiquitination mechanism.
CDK4/6 inhibition blocks cancer metastasis through a USP51-ZEB1-dependent deubiquitination mechanism.
Signal transduction and targeted therapy (2020-04-17)
Zhen Zhang, Jianjun Li, Yang Ou, Guang Yang, Kaiyuan Deng, Qiong Wang, Zhaoyang Wang, Wenhao Wang, Quansheng Zhang, Hang Wang, Wei Sun, Peiqing Sun, Shuang Yang
ABSTRACT
Tumor metastasis is the most common cause of cancer-related deaths, yet it remains poorly understood. The transcription factor zinc-finger E-box binding homeobox 1 (ZEB1) is involved in the epithelial-to-mesenchymal transition (EMT) and plays a pivotal role in tumor metastasis. However, the underlying mechanisms of the posttranslational modification of ZEB1 remain largely unknown. Herein, we demonstrated that specific inhibition of CDK4/6 was able to block tumor metastasis of breast cancer by destabilizing the ZEB1 protein in vitro and in vivo. Mechanistically, we determined that the deubiquitinase USP51 is a bona fide target of CDK4/6. The phosphorylation and activation of USP51 by CDK4/6 is necessary to deubiquitinate and stabilize ZEB1. Moreover, we found a strong positive correlation between the expression of p-RB (an indicator of CDK4/6 activity), p-USP51 and ZEB1 in metastatic human breast cancer samples. Notably, the high expression of p-RB, p-USP51, and ZEB1 was significantly correlated with a poor clinical outcome. Taken together, our results provide evidence that the CDK4/6-USP51-ZEB1 axis plays a key role in breast cancer metastasis and could be a viable therapeutic target for the treatment of advanced human cancers.
Sigma-Aldrich
ANTI-USP51 (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution