Merck
  • Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Cell (2020-12-06)
Timothy N Hoang, Maria Pino, Arun K Boddapati, Elise G Viox, Carly E Starke, Amit A Upadhyay, Sanjeev Gumber, Michael Nekorchuk, Kathleen Busman-Sahay, Zachary Strongin, Justin L Harper, Gregory K Tharp, Kathryn L Pellegrini, Shannon Kirejczyk, Keivan Zandi, Sijia Tao, Tristan R Horton, Elizabeth N Beagle, Ernestine A Mahar, Michelle Y H Lee, Joyce Cohen, Sherrie M Jean, Jennifer S Wood, Fawn Connor-Stroud, Rachelle L Stammen, Olivia M Delmas, Shelly Wang, Kimberly A Cooney, Michael N Sayegh, Lanfang Wang, Peter D Filev, Daniela Weiskopf, Guido Silvestri, Jesse Waggoner, Anne Piantadosi, Sudhir P Kasturi, Hilmi Al-Shakhshir, Susan P Ribeiro, Rafick P Sekaly, Rebecca D Levit, Jacob D Estes, Thomas H Vanderford, Raymond F Schinazi, Steven E Bosinger, Mirko Paiardini
ABSTRACT
SARS-CoV-2-induced hypercytokinemia and inflammation are critically associated with COVID-19 severity. Baricitinib, a clinically approved JAK1/JAK2 inhibitor, is currently being investigated in COVID-19 clinical trials. Here, we investigated the immunologic and virologic efficacy of baricitinib in a rhesus macaque model of SARS-CoV-2 infection. Viral shedding measured from nasal and throat swabs, bronchoalveolar lavages, and tissues was not reduced with baricitinib. Type I interferon (IFN) antiviral responses and SARS-CoV-2-specific T cell responses remained similar between the two groups. Animals treated with baricitinib showed reduced inflammation, decreased lung infiltration of inflammatory cells, reduced NETosis activity, and more limited lung pathology. Importantly, baricitinib-treated animals had a rapid and remarkably potent suppression of lung macrophage production of cytokines and chemokines responsible for inflammation and neutrophil recruitment. These data support a beneficial role for, and elucidate the immunological mechanisms underlying, the use of baricitinib as a frontline treatment for inflammation induced by SARS-CoV-2 infection.
Product Description
Roche
Red Blood Cell Lysis Buffer, solution, Roche, pkg of 100 mL, sufficient for 50-500 reactions
Sigma-Aldrich
Anti-MxA, clone M143 (CL143), clone CL143, from mouse