Merck
  • Kinetics of SARS-CoV-2 specific IgM and IgG responses in COVID-19 patients.
Kinetics of SARS-CoV-2 specific IgM and IgG responses in COVID-19 patients.
Emerging microbes & infections (2020-05-03)
Baoqing Sun, Ying Feng, Xiaoneng Mo, Peiyan Zheng, Qian Wang, Pingchao Li, Ping Peng, Xiaoqing Liu, Zhilong Chen, Huimin Huang, Fan Zhang, Wenting Luo, Xuefeng Niu, Peiyu Hu, Longyu Wang, Hui Peng, Zhifeng Huang, Liqiang Feng, Feng Li, Fuchun Zhang, Fang Li, Nanshan Zhong, Ling Chen
ABSTRACT
The emerging COVID-19 caused by SARS-CoV-2 infection poses severe challenges to global public health. Serum antibody testing is becoming one of the critical methods for the diagnosis of COVID-19 patients. We investigated IgM and IgG responses against SARS-CoV-2 nucleocapsid (N) and spike (S) protein after symptom onset in the intensive care unit (ICU) and non-ICU patients. 130 blood samples from 38 COVID-19 patients were collected. The levels of IgM and IgG specific to N and S protein were detected by ELISA. A series of blood samples were collected along the disease course from the same patient, including 11 ICU patients and 27 non-ICU patients for longitudinal analysis. N and S specific IgM and IgG (N-IgM, N-IgG, S-IgM, S-IgG) in non-ICU patients increased after symptom onset. N-IgM and S-IgM in some non-ICU patients reached a peak in the second week, while N-IgG and S-IgG continued to increase in the third week. The combined detection of N and S specific IgM and IgG could identify up to 75% of SARS-CoV-2 infected patients in the first week. S-IgG was significantly higher in non-ICU patients than in ICU patients in the third week. In contrast, N-IgG was significantly higher in ICU patients than in non-ICU patients. The increase of S-IgG positively correlated with the decrease of C-reactive protein (CRP) in non-ICU patients. N and S specific IgM and IgG increased gradually after symptom onset and can be used for detection of SARS-CoV-2 infection. Analysis of the dynamics of S-IgG may help to predict prognosis.
Sigma-Aldrich
Goat Anti-Human IgM Antibody, Fc5µ, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
Goat Anti-Human IgG Antibody, HRP conjugate, Chemicon®, from goat