Merck
  • Heat stress affects some physiological and productive variables and alters metabolism in dairy ewes.
Heat stress affects some physiological and productive variables and alters metabolism in dairy ewes.
Journal of dairy science (2020-11-10)
Nabil Mehaba, Wellington Coloma-Garcia, Xavier Such, Gerardo Caja, Ahmed A K Salama
ABSTRACT
Heat stress (HS) has a significant economic impact on the global dairy industry. However, the mechanisms by which HS negatively affects metabolism and milk synthesis in dairy ewes are not well defined. This study evaluated the production and metabolic variables in dairy ewes under controlled HS conditions. Eight Lacaune ewes (75.5 ± 3.2 kg of body weight; 165 ± 4 d of lactation; 2.31 ± 0.04 kg of milk per day) were submitted to thermoneutral (TN) or HS conditions in a crossover design (2 periods, 21 d each, 6-d transition). Conditions (day-night, 12-12 h; relative humidity; temperature-humidity index, THI) were: TN (15-20°C; 50 ± 5%; THI = 59-65) and HS (28-35°C; 45 ± 5%; THI = 75-83). Ewes were fed ad libitum and milked twice daily. Rectal temperature, respiratory rate, feed intake, water consumption, and milk yield were recorded daily. Milk and blood samples were collected weekly. Additionally, TN and HS ewes were exposed to glucose tolerance test, insulin tolerance test, and epinephrine challenge. Heat stress reduced feed intake (-11%), and increased rectal temperature (+0.77°C), respiratory rate (+90 breaths/min), and water consumption (+28%). Despite the reduced feed intake, HS ewes produced similar milk to TN ewes, but their milk contained lower fat (-1.7 points) and protein (-0.86 points). Further, HS milk tended to contain more somatic cells (+0.23 log points). Blood creatinine was greater in HS compared with TN, but no differences in blood glucose, nonesterified fatty acids, or urea were detected. When glucose was infused, TN and HS had similar insulin response, but higher glucose response (+85%) was detected in HS ewes. Epinephrine infusion resulted in lower nonesterified fatty acids response (-215%) in HS than TN ewes. Overall, HS decreased feed intake, but milk production was not affected. Heat stress caused metabolic adaptations that included increased body muscle degradation and reduced adipose tissue mobilization. These adaptations allowed ewes to spare glucose and to avoid reductions in milk yield.
Sigma-Aldrich
(−)-Epinephrine
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.