Merck
  • Resonant and Sensing Performance of Volume Waveguide Structures Based on Polymer Nanomaterials.
Resonant and Sensing Performance of Volume Waveguide Structures Based on Polymer Nanomaterials.
Nanomaterials (Basel, Switzerland) (2020-10-30)
Tatiana Smirnova, Volodymyr Fitio, Oksana Sakhno, Pavel Yezhov, Andrii Bendziak, Volodymyr Hryn, Stefano Bellucci
ABSTRACT
Organic-inorganic photocurable nanocomposite materials are a topic of intensive research nowadays. The wide variety of materials and flexibility of their characteristics provide more freedom to design optical elements for light and neutron optics and holographic sensors. We propose a new strategy of nanocomposite application for fabricating resonant waveguide structures (RWS), whose working principle is based on optical waveguide resonance. Due to their resonant properties, RWS can be used as active tunable filters, refractive index (RI) sensors, near-field enhancers for spectroscopy, non-linear optics, etc. Our original photocurable organic-inorganic nanocomposite was used as a material for RWS. Unlike known waveguide structures with corrugated surfaces, we investigated the waveguide gratings with the volume modulation of the RI fabricated by a holographic method that enables large-size structures with high homogeneity. In order to produce thin photosensitive waveguide layers for their subsequent holographic structuring, a special compression method was developed. The resonant and sensing properties of new resonant structures were experimentally examined. The volume waveguide gratings demonstrate narrow resonant peaks with a bandwidth less than 0.012 nm. The Q-factor exceeds 50,000. The sensor based on waveguide volume grating provides detection of a minimal RI change of 1 × 10-4 RIU. Here we also present the new theoretical model that is used for analysis and design of developed RWS. Based on the proposed model, fairly simple analytical relationships between the parameters characterizing the sensor were obtained.
Sigma-Aldrich
Isobutyl acrylate, ≥99%, contains 10-20 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.