Merck
  • RET Solvent Front Mutations Mediate Acquired Resistance to Selective RET Inhibition in RET-Driven Malignancies.
RET Solvent Front Mutations Mediate Acquired Resistance to Selective RET Inhibition in RET-Driven Malignancies.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2020-01-29)
Benjamin J Solomon, Lavinia Tan, Jessica J Lin, Stephen Q Wong, Sebastian Hollizeck, Kevin Ebata, Brian B Tuch, Satoshi Yoda, Justin F Gainor, Lecia V Sequist, Geoffrey R Oxnard, Oliver Gautschi, Alexander Drilon, Vivek Subbiah, Christine Khoo, Edward Y Zhu, Michele Nguyen, Dahlia Henry, Kevin R Condroski, Gabrielle R Kolakowski, Eliana Gomez, Joshua Ballard, Andrew T Metcalf, James F Blake, Sarah-Jane Dawson, Wayne Blosser, Louis F Stancato, Barbara J Brandhuber, Steve Andrews, Bruce G Robinson, S Michael Rothenberg
ABSTRACT
Novel rearranged in transfection (RET)-specific tyrosine kinase inhibitors (TKIs) such as selpercatinib (LOXO-292) have shown unprecedented efficacy in tumors positive for RET fusions or mutations, notably RET fusion-positive NSCLC and RET-mutated medullary thyroid cancer (MTC). However, the mechanisms of resistance to these agents have not yet been described. Analysis was performed of circulating tumor DNA and tissue in patients with RET fusion-positive NSCLC and RET-mutation positive MTC who developed disease progression after an initial response to selpercatinib. Acquired resistance was modeled preclinically using a CCDC6-RET fusion-positive NSCLC patient-derived xenograft. The inhibitory activity of anti-RET multikinase inhibitors and selective RET TKIs was evaluated in enzyme and cell-based assays. After a dramatic initial response to selpercatinib in a patient with KIF5B-RET NSCLC, analysis of circulating tumor DNA revealed emergence of RET G810R, G810S, and G810C mutations in the RET solvent front before the emergence of clinical resistance. Postmortem biopsy studies reported intratumor and intertumor heterogeneity with distinct disease subclones containing G810S, G810R, and G810C mutations in multiple disease sites indicative of convergent evolution on the G810 residue resulting in a common mechanism of resistance. Acquired mutations in RET G810 were identified in tumor tissue from a second patient with CCDC6-RET fusion-positive NSCLC and in plasma from patients with additional RET fusion-positive NSCLC and RET-mutant MTC progressing on an ongoing phase 1 and 2 trial of selpercatinib. Preclinical studies reported the presence of RET G810R mutations in a CCDC6-RET patient-derived xenograft (from a patient with NSCLC) model of acquired resistance to selpercatinib. Structural modeling predicted that these mutations sterically hinder the binding of selpercatinib, and in vitro assays confirmed loss of activity for both anti-RET multikinase inhibitors and selective RET TKIs. RET G810 solvent front mutations represent the first described recurrent mechanism of resistance to selective RET inhibition with selpercatinib. Development of potent inhibitor of these mutations and maintaining activity against RET gatekeeper mutations could be an effective strategy to target resistance to selective RET inhibitors.
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography