Merck
  • Discovery of Covalent CDK14 Inhibitors with Pan-TAIRE Family Specificity.
Discovery of Covalent CDK14 Inhibitors with Pan-TAIRE Family Specificity.
Cell chemical biology (2019-04-02)
Fleur M Ferguson, Zainab M Doctor, Scott B Ficarro, Christopher M Browne, Jarrod A Marto, Jared L Johnson, Tomer M Yaron, Lewis C Cantley, Nam Doo Kim, Taebo Sim, Matthew J Berberich, Marian Kalocsay, Peter K Sorger, Nathanael S Gray
ABSTRACT
Cyclin-dependent kinase 14 (CDK14) and other TAIRE family kinases (CDKs 15-18) are proteins that lack functional annotation but are frequent off-targets of clinical kinase inhibitors. In this study we develop and characterize FMF-04-159-2, a tool compound that specifically targets CDK14 covalently and possesses a TAIRE kinase-biased selectivity profile. This tool compound and its reversible analog were used to characterize the cellular consequences of covalent CDK14 inhibition, including an unbiased investigation using phospho-proteomics. To reduce confounding off-target activity, washout conditions were used to deconvolute CDK14-specific effects. This investigation suggested that CDK14 plays a supporting role in cell-cycle regulation, particularly mitotic progression, and identified putative CDK14 substrates. Together, these results represent an important step forward in understanding the cellular consequences of inhibiting CDK14 kinase activity.
Sigma-Aldrich
ANTI-PCTAIRE2 (PCTK2)(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2) Antibody, clone 3E10, clone 3E10, from rat