Merck
  • Obesity promotes gastric cancer metastasis via diacylglycerol acyltransferase 2-dependent lipid droplets accumulation and redox homeostasis.
Obesity promotes gastric cancer metastasis via diacylglycerol acyltransferase 2-dependent lipid droplets accumulation and redox homeostasis.
Redox biology (2020-06-09)
Shuai Li, Teng Wu, Yun-Xin Lu, Jin-Xiang Wang, Feng-Hai Yu, Mei-Zhu Yang, Yi-Jia Huang, Zhi-Jing Li, Sen-Lan Wang, Ling Huang, Lei Lu, Tian Tian
ABSTRACT
Experimental and molecular epidemiological studies indicate important roles for adipose tissue or high-fat diet (HFD) in tumor growth and metastasis. Gastric cancer (GC) possesses a metastatic predilection for the adipocyte-rich peritoneum. However, the precise molecular relevance of HFD in the peritoneal metastasis of GC remains unclear. Here, we showed that HFD causes obvious fat accumulation and promotes peritoneal dissemination of GC in vivo. Peritoneum-derived adipocytes induces robust lipid droplet (LD) accumulation and fatty acid oxidation in GC cells through transcriptional upregulation of DGAT2 in a C/EBPα-dependent manner and prevents anoikis during peritoneal dissemination. Treatment of GC cells with FAs or coculture with adipocytes induces intracellular formation of LDs and production of NADPH to overcome oxidative stress in vitro. Importantly, overexpression of DGAT2 was identified as an independent predictor of poor survival that promotes lung and peritoneal metastasis of GC, and genetic or pharmacological inhibition of DGAT2, via disruption of lipid droplet formation in a lipid-rich environment, enhances the sensitivity of GC to anoikis in vitro and inhibits peritoneal metastasis in vivo. Overall, our findings highlight the notion that DGAT2 may be a promising therapeutic target in GC with peritoneal implantation and provide some evidence for uncovering the link between obesity and tumor metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human DGAT2
Pricing and availability is not currently available.