Merck
  • Intratumoral Copper Modulates PD-L1 Expression and Influences Tumor Immune Evasion.
Intratumoral Copper Modulates PD-L1 Expression and Influences Tumor Immune Evasion.
Cancer research (2020-08-21)
Florida Voli, Emanuele Valli, Luigi Lerra, Kathleen Kimpton, Federica Saletta, Federico M Giorgi, Daniele Mercatelli, Jourdin R C Rouaen, Sylvie Shen, Jayne E Murray, Aria Ahmed-Cox, Giuseppe Cirillo, Chelsea Mayoh, Paul A Beavis, Michelle Haber, Joseph A Trapani, Maria Kavallaris, Orazio Vittorio
ABSTRACT
Therapeutic checkpoint antibodies blocking programmed death receptor 1/programmed death ligand 1 (PD-L1) signaling have radically improved clinical outcomes in cancer. However, the regulation of PD-L1 expression on tumor cells is still poorly understood. Here we show that intratumoral copper levels influence PD-L1 expression in cancer cells. Deep analysis of the The Cancer Genome Atlas database and tissue microarrays showed strong correlation between the major copper influx transporter copper transporter 1 (CTR-1) and PD-L1 expression across many cancers but not in corresponding normal tissues. Copper supplementation enhanced PD-L1 expression at mRNA and protein levels in cancer cells and RNA sequencing revealed that copper regulates key signaling pathways mediating PD-L1-driven cancer immune evasion. Conversely, copper chelators inhibited phosphorylation of STAT3 and EGFR and promoted ubiquitin-mediated degradation of PD-L1. Copper-chelating drugs also significantly increased the number of tumor-infiltrating CD8+ T and natural killer cells, slowed tumor growth, and improved mouse survival. Overall, this study reveals an important role for copper in regulating PD-L1 and suggests that anticancer immunotherapy might be enhanced by pharmacologically reducing intratumor copper levels. SIGNIFICANCE: These findings characterize the role of copper in modulating PD-L1 expression and contributing to cancer immune evasion, highlighting the potential for repurposing copper chelators as enhancers of antitumor immunity. GRAPHICAL ABSTRACT: http://cancerres.aacrjournals.org/content/canres/80/19/4129/F1.large.jpg.
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Tetraethylenepentamine pentahydrochloride, 98%
Sigma-Aldrich
Gefitinib, ≥98% (HPLC)