Merck
Modulation of human immune responses by bovine interleukin-10.
PloS one (2011-04-06)
Gerco den Hartog, Huub F J Savelkoul, Ruud Schoemaker, Edwin Tijhaar, Adrie H Westphal, Talitha de Ruiter, Elise van de Weg-Schrijver, R J Joost van Neerven
ABSTRACT
Cytokines can be functionally active across species barriers. Bovine IL-10 has an amino acid sequence identity with human IL-10 of 76.8%. Therefore, the aim of this study was to evaluate whether bovine IL-10 has immunomodulatory activities on human monocytes and dendritic cells. Peripheral blood monocytes were isolated from healthy donors, and used directly or allowed to differentiate to dendritic cells under the influence of IL-4 and GM-CSF. Recombinant bovine IL-10 inhibited TLR induced activation of monocytes, and dose-dependently inhibited LPS-induced activation of monocyte-derived DCs comparable to human IL-10. By using blocking antibodies to either bovine IL-10 or the human IL-10 receptor it was demonstrated that inhibition of monocyte activation by bovine IL-10 was dependent on binding of bovine IL-10 to the human IL-10R. These data demonstrate that bovine IL-10 potently inhibits the activation of human myeloid cells in response to TLR activation. Bovine IL-10 present in dairy products may thus potentially contribute to the prevention of necrotizing enterocolitis and allergy, enhance mucosal tolerance induction and decrease intestinal inflammation and may therefore be applicable in infant foods and in immunomodulatory diets.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, BioXtra, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, purified by ion-exchange chromatography
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by gel-filtration chromatography
Sigma-Aldrich
Peptidoglycan from Staphylococcus aureus, cell wall component