Merck
  • Comparison between a Conventional Anti-Biofouling Compound and a Novel Modified Low-Fouling Polyethersulfone Ultrafiltration Membrane: Bacterial Anti-Attachment, Water Quality and Productivity.
Comparison between a Conventional Anti-Biofouling Compound and a Novel Modified Low-Fouling Polyethersulfone Ultrafiltration Membrane: Bacterial Anti-Attachment, Water Quality and Productivity.
Membranes (2020-09-16)
Norhan Nady, Noha Salem, Ranya Amer, Ahmed El-Shazly, Sherif H Kandil, Mohamed Salah El-Din Hassouna
ABSTRACT
In this work, the efficiency of a conventional chlorination pretreatment is compared with a novel modified low-fouling polyethersulfone (PES) ultrafiltration (UF) membrane, in terms of bacteria attachment and membrane biofouling reduction. This study highlights the use of membrane modification as an effective strategy to reduce bacterial attachment, which is the initial step of biofilm formation, rather than using antimicrobial agents that can enhance bacterial regrowth. The obtained results revealed that the filtration of pretreated, inoculated seawater using the modified PES UF membrane without the pre-chlorination step maintained the highest initial flux (3.27 ± 0.13 m3·m-2·h-1) in the membrane, as well as having one and a half times higher water productivity than the unmodified membrane. The highest removal of bacterial cells was achieved by the modified membrane without chlorination, in which about 12.07 × 104 and 8.9 × 104 colony-forming unit (CFU) m-2 bacterial cells were retained on the unmodified and modified membrane surfaces, respectively, while 29.4 × 106 and 0.42 × 106 CFU mL-1 reached the filtrate for the unmodified and modified membranes, respectively. The use of chlorine disinfectant resulted in significant bacterial regrowth.
Sigma-Aldrich
N,N-Diethyl-p-phenylenediamine, 97%