Merck
  • The tRNA pseudouridine synthase TruB1 regulates the maturation of let-7 miRNA.
The tRNA pseudouridine synthase TruB1 regulates the maturation of let-7 miRNA.
The EMBO journal (2020-09-15)
Ryota Kurimoto, Tomoki Chiba, Yoshiaki Ito, Takahide Matsushima, Yuki Yano, Kohei Miyata, Yuka Yashiro, Tsutomu Suzuki, Kozo Tomita, Hiroshi Asahara
ABSTRACT
Let-7 is an evolutionary conserved microRNA that mediates post-transcriptional gene silencing to regulate a wide range of biological processes, including development, differentiation, and tumor suppression. Let-7 biogenesis is tightly regulated by several RNA-binding proteins, including Lin28A/B, which represses let-7 maturation. To identify new regulators of let-7, we devised a cell-based functional screen of RNA-binding proteins using a let-7 sensor luciferase reporter and identified the tRNA pseudouridine synthase, TruB1. TruB1 enhanced maturation specifically of let-7 family members. Rather than inducing pseudouridylation of the miRNAs, high-throughput sequencing crosslinking immunoprecipitation (HITS-CLIP) and biochemical analyses revealed direct binding between endogenous TruB1 and the stem-loop structure of pri-let-7, which also binds Lin28A/B. TruB1 selectively enhanced the interaction between pri-let-7 and the microprocessor DGCR8, which mediates miRNA maturation. Finally, TruB1 suppressed cell proliferation, which was mediated in part by let-7. Altogether, we reveal an unexpected function for TruB1 in promoting let-7 maturation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Pseudouridine, ≥98% (HPLC)
Pricing and availability is not currently available.