Merck
  • Biological activity of astilbin from Dimorphandra mollis against Anticarsia gemmatalis and Spodoptera frugiperda.
Biological activity of astilbin from Dimorphandra mollis against Anticarsia gemmatalis and Spodoptera frugiperda.
Pest management science (2002-05-10)
Luciane G Batista Pereira, Fernando Petacci, João B Fernandes, Arlene G Corrêa, Paulo C Vieira, M Fátima G F da Silva, Osmar Malaspina
ABSTRACT
Astilbin was isolated in high yield from Dimorphandra mollis, and its insecticidal and growth inhibiting activity by stomach ingestion were evaluated against Anticarsia gemmatalis and Spodoptera frugiperda. The insecticidal activity of astilbin, the weight reduction of the larval phase and the prolongation of the larval and pupal phases were verified for both species. Astilbin was identified on the base of its NMR, MS and physical data.
Sigma-Aldrich
Astilbin from Engelhardtia roxburghiana, ≥98%
Pricing and availability is not currently available.