Merck
  • [Study on antidepressant components of sucrose ester from Polygala tenuifolia].
[Study on antidepressant components of sucrose ester from Polygala tenuifolia].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-10-04)
Hai-Hua Tu, Ping Liu, Liang Mu, Hong-Bo Liao, Ting-Ting Xie, Li-Hua Ma, Yao-Ming Liu
ABSTRACT
To investigate the antidepressant components of Polygala tenuifolia. The chromatographic method was used to isolate and purify the chemical constituents, their structures were identified by spectral analysis, MTT method was applied to investigate their cytotoxic activities. Nine compounds were isolated from the roots of P. tenuifolia. Their structures were identified as sibiricose A, (1), sibiricose A5 (2), tenuifoliside A (3) and 3', 6-disinapoyl sucrose (4), sibiricose A6 (5), 3, 4, 5-trimethoxycinnamate (6), polygalaxanthone III (7), tenuifolioses A (8), tenuifolioses H (9) and some compounds' activities to PC12 were observed. Compound 1 was isolated from this plant for the first time. Compounds 2,3 could protect PC12 cells damage induced by P. tenuifolia.
Sigma-Aldrich
Sibiricose A5, ≥85% (LC/MS-ELSD)
Pricing and availability is not currently available.