Merck
  • Association between MMP/TIMP Levels in the Aqueous Humor and Plasma with Axial Lengths in Myopia Patients.
Association between MMP/TIMP Levels in the Aqueous Humor and Plasma with Axial Lengths in Myopia Patients.
BioMed research international (2020-07-01)
Yu Yue, Yu-Wen Hsiao, Ji-Bo Zhou
ABSTRACT
The present study investigated the profiles of matrix metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) of the aqueous humor (AH) and plasma (PL) in myopia patients, to determine whether there was an association between these levels with their axial length (AL) and to investigate if MMPs/TIMPs were regulated locally or systemically. A cross-sectional study was conducted. Thirty-nine patients (78 eyes) diagnosed with high myopia were recruited. The AL was measured using IOL Master. And the patients were divided into three groups based on their AL, Group A (AL ≤ 26 mm), Group B (26 < AL ≤ 28 mm), and Group C (AL > 28 mm). The AH in both eyes and blood samples were collected before the patients underwent implantable collamer lens surgery. In all, 78 samples of the AH and 39 samples of the PL were analyzed using MILLIPLEX map assays, followed by statistical analyses of the results. There were 8 patients (16 eyes) in Group A, 22 patients (44 eyes) in Group B, and 9 patients (18 eyes) in Group C. MMP-1 (p = 0.014, Β = 0.118), MMP-2 (p ≤ 0.001, Β = 0.278), MMP-9 (p ≤ 0.001, Β = 0.019), and TIMP-1 (p = 0.014, Β = 0.062) in the AH were positively associated with the AL. MMP-1 (p = 0.004, Β = 0.001) and TIMP-1 (p = 0.030, Β = 1.171) concentrations in the PL increased linearly with longer ALs. No concentration-dependent relationship was found between MMP-2 in the PL and AL. There was a consistent relationship between MMP-2 in the AH and AL. AL was not consistently or substantially affected by MMP-2 in the PL, indicating myopia formation was possibly a localized process. Associations among MMP-1, MMP-9, and TIMP-1 in the AH and AL were also observed.