Merck
  • Knockdown of MCM10 Gene Impairs Glioblastoma Cell Proliferation, Migration and Invasion and the Implications for the Regulation of Tumorigenesis.
Knockdown of MCM10 Gene Impairs Glioblastoma Cell Proliferation, Migration and Invasion and the Implications for the Regulation of Tumorigenesis.
Journal of molecular neuroscience : MN (2020-02-08)
Peng Kang, Zhe Han, Zhiyi Liao, Heng Zhang, Wang Jia, Yongji Tian
ABSTRACT
Minichromosome maintenance 10 (MCM10) plays an important role in DNA replication and is expressed in a variety of tumors, including glioma. However, its role and mechanism in glioma remain elusive. The purpose of this study was to examine the molecular function of MCM10 in glioblastoma cell lines in vitro and to further investigate the molecular mechanisms in the network mediated by MCM10. Cell proliferation, invasion, and migration were investigated in the absence of MCM10 mediated by RNA interference (RNAi) in U87 and U251 cell lines. Microarray data were obtained from U87 cells infected with a lentivirus expressing a small interfering RNA (siRNA) targeting MCM10, and ingenuity pathway analysis (IPA) was performed. Molecular signaling pathways, gene functions, and upstream and downstream regulatory genes and networks were analyzed. MCM10 was positively stained in human glioblastoma multiforme (GBM) samples according to immunohistochemistry. Silencing MCM10 in U87 and U251 cells significantly reduced cell proliferation, migration, and invasion. In U87 cells transfected with MCM10, 274 genes were significantly upregulated, while 313 genes were downregulated. IPA revealed that MCM10 is involved in the IGF-1 signaling pathway, and calcitriol appears to be a significant upstream regulator of MCM10. Other factors, such as TWIST1 and Stat3, also interact within the MCM10-mediated network. Our data indicate that MCM10 is involved in the regulation of GBM in vitro and may provide more evidence for understanding the molecular mechanisms of this fatal disease.
Sigma-Aldrich
MISSION® esiRNA, targeting human MCM10