Merck
  • Spaced Training Forms Complementary Long-Term Memories of Opposite Valence in Drosophila.
Spaced Training Forms Complementary Long-Term Memories of Opposite Valence in Drosophila.
Neuron (2020-04-15)
Pedro F Jacob, Scott Waddell
ABSTRACT
Forming long-term memory (LTM) often requires repetitive experience spread over time. Studies in Drosophila suggest aversive olfactory LTM is optimal after spaced training, multiple trials of differential odor conditioning with rest intervals. Memory after spaced training is frequently compared to that after the same number of trials without intervals. Here we show that, after spaced training, flies acquire additional information and form an aversive memory for the shock-paired odor and a slowly emerging and more persistent "safety-memory" for the explicitly unpaired odor. Safety-memory acquisition requires repetition, order, and spacing of the training trials and relies on triggering specific rewarding dopaminergic neurons. Co-existence of aversive and safety memories is evident as depression of odor-specific responses at different combinations of junctions in the mushroom body output network; combining two outputs appears to signal relative safety. Having complementary aversive and safety memories augments LTM performance after spaced training by making the odor preference more certain.
Sigma-Aldrich
3-Octanol, 99%
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%
Sigma-Aldrich
4-Methylcyclohexanol, mixture of cis and trans, 98%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)