Merck
  • Activation of Inward Rectifier K+ Channel 2.1 by PDGF-BB in Rat Vascular Smooth Muscle Cells through Protein Kinase A.
Activation of Inward Rectifier K+ Channel 2.1 by PDGF-BB in Rat Vascular Smooth Muscle Cells through Protein Kinase A.
BioMed research international (2020-05-29)
Chengchun Tang, Dong Wang, Erfei Luo, Gaoliang Yan, Bo Liu, Jiantong Hou, Yong Qiao
ABSTRACT
Platelet-derived growth factor-BB (PDGF-BB) can induce the proliferation, migration, and phenotypic modulation of vascular smooth muscle cells (VSMCs). We used patch clamp methods to study the effects of PDGF-BB on inward rectifier K+ channel 2.1 (Kir2.1) channels in rat thoracic aorta VSMCs (RASMCs). PDGF-BB (25 ng/mL) increased Kir2.x currents (-11.81 ± 2.47 pA/pF, P < 0.05 vs. CON, n = 10). Ba2+(50 μM) decreased Kir2.x currents (-2.13 ± 0.23 pA/pF, P < 0.05 vs. CON, n = 10), which were promoted by PDGF-BB (-6.98 ± 1.03 pA/pF). PDGF-BB specifically activates Kir2.1 but not Kir2.2 and Kir2.3 channels in HEK-293 cells. The PDGF-BB-induced stimulation of Kir2.1 currents was blocked by the PDGF-BB receptor β (PDGF-BBRβ) inhibitor AG1295 and was not affected by the PDGF-BBRα inhibitor AG1296. The PDGF-BB-induced stimulation of Kir2.1 currents was blocked by the protein kinase A inhibitor Rp-8-CPT-cAMPs; however, the antagonist of protein kinase B (GSK690693) had marginal effects on current activity. The PDGF-BB-induced stimulation of Kir2.1 currents was enhanced by forskolin, an adenylyl cyclase (AC) activator, and was blocked by the AC inhibitor SQ22536. We conclude that PDGF-BB increases Kir2.1 currents via PDGF-BBRβ through activation of cAMP-PKA signaling in RASMCs.
Sigma-Aldrich
GSK690693, ≥98% (HPLC)