Merck
  • Matrix Isolation Spectroscopy and Nuclear Spin Conversion of Propyne Suspended in Solid Parahydrogen.
Matrix Isolation Spectroscopy and Nuclear Spin Conversion of Propyne Suspended in Solid Parahydrogen.
The journal of physical chemistry. A (2020-05-14)
A I Strom, A Gutiérrez-Quintanilla, M Chevalier, J Ceponkus, C Crépin, D T Anderson
ABSTRACT
Parahydrogen (pH2) quantum solids are excellent matrix isolation hosts for studying the rovibrational dynamics and nuclear spin conversion (NSC) kinetics of molecules containing indistinguishable nuclei with nonzero spin. The relatively slow NSC kinetics of propyne (CH3CCH) isolated in solid pH2 is employed as a tool to assign the rovibrational spectrum of propyne in the 600-7000 cm-1 region. Detailed analyses of a variety of parallel (ΔK = 0) and perpendicular (ΔK=±1) bands of propyne indicate that the end-over-end rotation of propyne is quenched, but K rotation of the methyl group around the C3 symmetry axis still persists. However, this single-axis K rotation is significantly hindered for propyne trapped in solid pH2 such that the energies of the K rotational states do not obey simple energy-level expressions. The NSC kinetics of propyne follows first-order reversible kinetics with a 287(7) min effective time constant at 1.7 K. Intensity-intensity correlation plots are used to determine the relative line strengths of individual ortho- and para-propyne rovibrational transitions, enabling an independent estimation of the ground vibrational state effective A″ constant of propyne.
Sigma-Aldrich
Propyne, ≥99%