Merck
  • Dynamics of HSPA1A and redox status in the spermatozoa and fluid from different segments of goat epididymis.
Dynamics of HSPA1A and redox status in the spermatozoa and fluid from different segments of goat epididymis.
Cell stress & chaperones (2020-03-25)
Akhilesh Kumar, Brijesh Yadav, Dilip Kumar Swain, Mukul Anand, Arun Kumar Madan, Raj Kumar Singh Yadav, Bhawna Kushawaha, Sarvajeet Yadav
ABSTRACT
The present study was attempted to investigate the dynamics of HSPA1A and redox status in the spermatozoa and fluid of different segments of buck epididymis. Testes were collected from sexually mature and healthy bucks aged between 2 and 3 years. The fluid and spermatozoa from different segments (caput, corpus and cauda) were harvested for further processing and analysis. The concentration of HSPA1A in spermatozoa lysate and epididymal fluid and its relative mRNA expression in spermatozoa from different segments of epididymis were studied. The HSPA1A concentration in epididymal fluid was significantly (P < 0.01) higher in the corpus as compared with caput and cauda, whereas, its concentration and relative mRNA expression decreased significantly (P < 0.01) in the spermatozoa from caput to cauda. The activities of SOD, GR, GST, and concentrations of manoldialdehyde and ROS decreased significantly (P < 0.01) in the spermatozoa from caput to cauda. The glutathione concentration and GPx activity decreased significantly (P < 0.01) in the spermatozoa of cauda as compared with the corpus. The SOD activity and ROS concentration were significantly (P < 0.01) higher in corpus, and GR and GST activity were significantly (P < 0.01) higher in caput fluid as compared with corpus and cauda. It may be concluded that HSPA1A concentration and its relative mRNA expression in spermatozoa decreased progressively, and redox status was altered during transit from caput to cauda.
Sigma-Aldrich
Human Heat Shock Protein 27 ELISA Kit, for serum, plasma, cell culture supernatant and urine