Merck
  • Grandmaternal betaine supplementation enhances hepatic IGF2 expression in F2 rat offspring through modification of promoter DNA methylation.
Grandmaternal betaine supplementation enhances hepatic IGF2 expression in F2 rat offspring through modification of promoter DNA methylation.
Journal of the science of food and agriculture (2019-11-23)
Shu Yang, Nannan Zhao, Bo Sun, Yang Yang, Yun Hu, Ruqian Zhao
ABSTRACT
We reported previously that maternal betaine promotes hepatic insulin-like growth factor (IGF2) expression in F1 offspring rats through hypermethylation of the IGF2/H19 imprinting control region (ICR). It remains unknown whether this acquired trait can be transmitted to the F2 generation. This study aimed to determine whether dietary betaine supplementation to grand dams affects the hepatic IGF2 expression in F2 rat offspring and how it is related to alterations in DNA methylation. F2 rat offspring derived from grand dams fed basal or betaine-supplemented diet (10 g kg-1 ) were examined at weaning. Serum IGF2 concentration was measured with enzyme-linked immunosorbent assay (ELISA). Hepatic expression of IGF2, together with other proliferation and apoptosis markers, was determined by using quantitative polymerase chain reaction (qPCR), western blot, and immunohistochemistry. The methylation status of the IGF2/H19 ICR and the promoters of IGF2 gene were detected by methylated DNA immunoprecipitation quantitative polymerase chain reaction (MeDIP-qPCR). The maternal betaine-induced up-regulation of hepatic IGF2 expression in F1 rat offspring was transmitted to the F2 generation. The F2 rats from the betaine group demonstrated enhanced hepatic IGF2 expression at both mRNA and protein levels, in association with higher serum IGF2 concentration. No alterations were observed in the ICR methylation of the IGF2/H19 locus, and hypomethylation was detected in promoters of IGF2 gene in betaine group. These results indicate that maternal betaine enhances hepatic IGF2 expression in F2 rat offspring through modification of DNA methylation on IGF2 promoters. © 2019 Society of Chemical Industry.
Sigma-Aldrich
Betaine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Pricing and availability is not currently available.