Merck
  • Effects of Chemical Modifications on the Thermoresponsive Behavior of a PDMAEA-b-PNIPAM-b-POEGA Triblock Terpolymer.
Effects of Chemical Modifications on the Thermoresponsive Behavior of a PDMAEA-b-PNIPAM-b-POEGA Triblock Terpolymer.
Polymers (2020-06-25)
Despoina Giaouzi, Stergios Pispas
ABSTRACT
In this work, the synthesis, selective chemical modifications, and self-assembly behavior in aqueous media of a novel poly(2-(dimethylamino)ethyl acrylate)20-b-poly(N-isopropylacrylamide)11-b-poly(oligo ethylene glycol methyl ether acrylate)18 (PDMAEA20-b-PNIPAM11-b-POEGA18) dual-responsive (pH and temperature) and triply hydrophilic amino-based triblock terpolymer are reported. The amine functional triblock terpolymer was synthesized by sequential reversible addition fragmentation chain transfer polymerization (RAFT) polymerization and molecularly characterized by size exclusion chromatography (SEC) and 1H-NMR spectroscopy that evidenced the success of the three-step polymerization scheme. The tertiary amine pendant groups of the PDMAEA block were chemically modified in order to produce the Q1PDMAEA20-b-PNIPAM11-b-POEGA18 as well as the Q6PDMAEA20-b-PNIPAM11-b-POEGA18 quaternized triblock terpolymers (Q1 and Q6 prefixes show the number of carbon atoms (C1 and C6) attached on the PDMAEA groups) using methyl iodide (CH3I) and 1-iodohexane (C6H13I) as the quaternizing agents and the SPDMAEA20-b-PNIPAM11-b-POEGA18 sulfobetainized triblock terpolymer using 1,3 propanesultone (C3H6O3S) as the sulfobetainization agent. The self-assembly properties of the triblock terpolymers in aqueous solutions upon varying temperature and solution pH were studied by light scattering and fluorescence spectroscopy experiments. The novel triblock terpolymers self-assemble into nanosized aggregates upon solution temperature rise above the nominal lower critical solution temperature (LCST) of the temperature-responsive PNIPAM block. The remarkable stimuli-responsive self-assembly behavior of the novel triblock terpolymers in aqueous media make them interesting candidates for biomedical applications in the fields of drug and gene delivery.
Sigma-Aldrich
2-(Dimethylamino)ethyl acrylate, contains <2,000 ppm MEHQ as inhibitor, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid, 98% (HPLC)
Pricing and availability is not currently available.