Merck
  • Antitumor activity of BIBF 1120, a triple angiokinase inhibitor, and use of VEGFR2+pTyr+ peripheral blood leukocytes as a pharmacodynamic biomarker in vivo.
Antitumor activity of BIBF 1120, a triple angiokinase inhibitor, and use of VEGFR2+pTyr+ peripheral blood leukocytes as a pharmacodynamic biomarker in vivo.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-12-07)
Kanae Kudo, Tokuzo Arao, Kaoru Tanaka, Tomoyuki Nagai, Kazuyuki Furuta, Kazuko Sakai, Hiroyasu Kaneda, Kazuko Matsumoto, Daisuke Tamura, Keiichi Aomatsu, Marco A De Velasco, Yoshihiko Fujita, Nagahiro Saijo, Masatoshi Kudo, Kazuto Nishio
ABSTRACT
BIBF 1120 is a potent, orally available triple angiokinase inhibitor that inhibits VEGF receptors (VEGFR) 1, 2, and 3, fibroblast growth factor receptors, and platelet-derived growth factor receptors. This study examined the antitumor effects of BIBF 1120 on hepatocellular carcinoma (HCC) and attempted to identify a pharmacodynamic biomarker for use in early clinical trials. We evaluated the antitumor and antiangiogenic effects of BIBF 1120 against HCC cell line both in vitro and in vivo. For the pharmacodynamic study, the phosphorylation levels of VEGFR2 in VEGF-stimulated peripheral blood leukocytes (PBL) were evaluated in mice inoculated with HCC cells and treated with BIBF 1120. BIBF 1120 (0.01 μmol/L) clearly inhibited the VEGFR2 signaling in vitro. The direct growth inhibitory effects of BIBF 1120 on four HCC cell lines were relatively mild in vitro (IC(50) values: 2-5 μmol/L); however, the oral administration of BIBF 1120 (50 or 100 mg/kg/d) significantly inhibited the tumor growth and angiogenesis in a HepG2 xenograft model. A flow cytometric analysis revealed that BIBF 1120 significantly decreased the phosphotyrosine (pTyr) levels of VEGFR2(+)CD45(dim) PBLs and the percentage of VEGFR2(+)pTyr(+) PBLs in vivo; the latter parameter seemed to be a more feasible pharmacodynamic biomarker. We found that BIBF 1120 exhibited potent antitumor and antiangiogenic activity against HCC and identified VEGFR2(+)pTyr(+) PBLs as a feasible and noninvasive pharmacodynamic biomarker in vivo.
Sigma-Aldrich
Nintedanib ethanesulphonate, ≥98% (HPLC)