Merck
  • Intramolecular chaperone-mediated secretion of an Rhs effector toxin by a type VI secretion system.
Intramolecular chaperone-mediated secretion of an Rhs effector toxin by a type VI secretion system.
Nature communications (2020-04-22)
Tong-Tong Pei, Hao Li, Xiaoye Liang, Zeng-Hang Wang, Guangfeng Liu, Li-Li Wu, Haeun Kim, Zhiping Xie, Ming Yu, Shuangjun Lin, Ping Xu, Tao G Dong
ABSTRACT
Bacterial Rhs proteins containing toxic domains are often secreted by type VI secretion systems (T6SSs) through unclear mechanisms. Here, we show that the T6SS Rhs-family effector TseI of Aeromonas dhakensis is subject to self-cleavage at both the N- and the C-terminus, releasing the middle Rhs core and two VgrG-interacting domains (which we name VIRN and VIRC). VIRC is an endonuclease, and the immunity protein TsiI protects against VIRC toxicity through direct interaction. Proteolytic release of VIRC and VIRN is mediated, respectively, by an internal aspartic protease activity and by two conserved glutamic residues in the Rhs core. Mutations abolishing self-cleavage do not block secretion, but reduce TseI toxicity. Deletion of VIRN or the Rhs core abolishes secretion. TseI homologs from Pseudomonas syringae, P. aeruginosa, and Vibrio parahaemolyticus are also self-cleaved. VIRN and VIRC interact with protein VgrG1, while the Rhs core interacts with protein TecI. We propose that VIRN and the Rhs core act as T6SS intramolecular chaperones to facilitate toxin secretion and function.
Sigma-Aldrich
Monoclonal Anti-6X His IgG, CF770 antibody produced in mouse, ~2 mg/mL, clone 33D10.D2.G8, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)