Merck
Neutrophil proteases degrade autoepitopes of NET-associated proteins.
Clinical and experimental immunology (2019-10-30)
C M de Bont, N Eerden, W C Boelens, G J M Pruijn
ABSTRACT
Neutrophils can form neutrophil extracellular traps (NETs) to capture microbes and facilitate their clearance. NETs consist of decondensed chromatin decorated with anti-microbial proteins. Here, we describe the effect of neutrophil proteases on the protein content of NETs. We show that the neutrophil serine proteases degrade several neutrophil proteins associated with NETs. Interestingly, the anti-bacterial proteins associated with NETs, such as myeloperoxidase, calgranulin B and neutrophil elastase (NE), seem to be less susceptible to proteolytic degradation than other NET proteins, such as actin and MNDA. NETs have been proposed to play a role in autoimmune reactions. Our data demonstrate that a large number of the autoepitopes of NET proteins that are recognized by autoantibodies produced by systemic lupus erythematosus (SLE) and rheumatoid arthritis (RA) patients are also removed by the proteases. In conclusion, neutrophil serine proteases have a major impact on the NET proteome and the proteolytic changes of NET-associated proteins may counteract autoimmune reactions to NET components.
Sigma-Aldrich
α1-Antitrypsin from human plasma, salt-free, lyophilized powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)