Merck
Diels-Alder reactions of cyclic isoimidium salts.
Organic letters (2010-09-17)
Robert K Boeckman, Yan Miller, Todd R Ryder
ABSTRACT
Diels-Alder reactions of cyclic isoimidium salts are described. The corresponding cycloadducts are obtained with high regio- and stereoselectivity. The use of homochiral cyclic isoimidium salts delivers cycloadducts with excellent diastereoselectivity (>99:1) that can be efficiently converted to enantiomerically pure lactones.
Sigma-Aldrich
(+)-Bis[(R)-1-phenylethyl]amine, 99%