Merck
  • Synthesis of dimethylformamide from CO2, H2 and dimethylamine over Cu/ZnO.
Synthesis of dimethylformamide from CO2, H2 and dimethylamine over Cu/ZnO.
Chemical communications (Cambridge, England) (2010-07-10)
Jinli Liu, Chengkun Guo, Zhaofu Zhang, Tao Jiang, Huizhen Liu, Jinliang Song, Honglei Fan, Buxing Han
ABSTRACT
Cheap and simple Cu/ZnO catalysts are very effective and recyclable for the synthesis of dimethylformamide (DMF) from CO(2), H(2), and dimethylamine, and a yield of 97% can be reached.