Merck
  • Incontinentia pigmenti: Generation of an IKBKG deficient human iPSC line (KICRi002-A-1) on a 46,XY background using CRISPR/Cas9.
Incontinentia pigmenti: Generation of an IKBKG deficient human iPSC line (KICRi002-A-1) on a 46,XY background using CRISPR/Cas9.
Stem cell research (2020-03-04)
Ambrin Fatima, Jens Schuster, Talia Akram, Carolina Maya González, Maria Sobol, Jan Hoeber, Niklas Dahl
ABSTRACT
Incontinentia pigmenti (IP) is an X-linked dominant neuroectodermal dysplasia caused by loss-of-function mutations in the IKBKG gene. Using CRISPR/Cas9 technology, we generated an IKBKG knock-out iPSC line (KICRi002-A-1) on a 46,XY background. The iPSC line showed a normal karyotype, expressed pluripotency markers and exhibited capability to differentiate into the three germ layers in vitro. Off-target editing was excluded and no IKBKG mRNA expression could be detected. Our line offers a useful resource to elucidate mechanisms caused by IKBKG deficiency that leads to disrupted male fetal development and for drug screening to improve treatment of female patients with IP.
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse