Merck
  • Pan-active imidazolopiperazine antimalarials target the Plasmodium falciparum intracellular secretory pathway.
Pan-active imidazolopiperazine antimalarials target the Plasmodium falciparum intracellular secretory pathway.
Nature communications (2020-04-15)
Gregory M LaMonte, Frances Rocamora, Danushka S Marapana, Nina F Gnädig, Sabine Ottilie, Madeline R Luth, Tilla S Worgall, Gregory M Goldgof, Roxanne Mohunlal, T R Santha Kumar, Jennifer K Thompson, Edgar Vigil, Jennifer Yang, Dylan Hutson, Trevor Johnson, Jianbo Huang, Roy M Williams, Bing Yu Zou, Andrea L Cheung, Prianka Kumar, Timothy J Egan, Marcus C S Lee, Dionicio Siegel, Alan F Cowman, David A Fidock, Elizabeth A Winzeler
ABSTRACT
A promising new compound class for treating human malaria is the imidazolopiperazines (IZP) class. IZP compounds KAF156 (Ganaplacide) and GNF179 are effective against Plasmodium symptomatic asexual blood-stage infections, and are able to prevent transmission and block infection in animal models. But despite the identification of resistance mechanisms in P. falciparum, the mode of action of IZPs remains unknown. To investigate, we here combine in vitro evolution and genome analysis in Saccharomyces cerevisiae with molecular, metabolomic, and chemogenomic methods in P. falciparum. Our findings reveal that IZP-resistant S. cerevisiae clones carry mutations in genes involved in Endoplasmic Reticulum (ER)-based lipid homeostasis and autophagy. In Plasmodium, IZPs inhibit protein trafficking, block the establishment of new permeation pathways, and cause ER expansion. Our data highlight a mechanism for blocking parasite development that is distinct from those of standard compounds used to treat malaria, and demonstrate the potential of IZPs for studying ER-dependent protein processing.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)