Merck
  • Upregulation of heme oxygenase-1 by Brahma-related gene 1 through Nrf2 signaling confers protective effect against high glucose-induced oxidative damage of retinal ganglion cells.
Upregulation of heme oxygenase-1 by Brahma-related gene 1 through Nrf2 signaling confers protective effect against high glucose-induced oxidative damage of retinal ganglion cells.
European journal of pharmacology (2020-02-28)
Wentao Sun, Jingni Yu, Qianyan Kang
ABSTRACT
High glucose (HG)-induced oxidative damage of retinal ganglion cells (RGCs) contributes to the pathogenesis of diabetic retinopathy, a severe complication of diabetes mellitus. Brahma-related gene 1 (Brg1) has currently emerged as a cytoprotective protein that alleviates oxidative damage induced by various stress. However, whether Brg1 is involved in the regulation of HG-induced oxidative damage of RGCs remains unknown. In this study, we aimed to investigate the potential role and underlying mechanism of Brg1 in regulating HG-induced damage of RGCs. We found that Brg1 expression was significantly downregulated in RGCs in response to HG treatment. Functional experiments showed that Brg1 knockdown enhanced HG-induced apoptosis and production of reactive oxygen species, while Brg1 overexpression suppressed HG-induced apoptosis and reactive oxygen species production, showing a protective effect. Moreover, Brg1 overexpression resulted in an increase in nuclear expression of nuclear factor-erythroid-2-related factor-2 (Nrf2) and the expression of heme oxygenase-1 (HO-1) in RGCs. Notably, inhibition of Nrf2 or HO-1 significantly blocked Brg1-mediated protection against HG-induced damage. Overall, these findings demonstrate that Brg1 protects RGCs from HG-induced oxidative damage through promotion of Nrf2/HO-1 signaling, indicating a potential role of Brg1 in the pathogenesis of diabetic retinopathy.
Sigma-Aldrich
MISSION® esiRNA, targeting human SMARCA4
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder