Merck
  • Glycine receptor α4 subunit facilitates the early embryonic development in mice.
Glycine receptor α4 subunit facilitates the early embryonic development in mice.
Reproduction (Cambridge, England) (2019-11-07)
Hirofumi Nishizono, Mohamed Darwish, Takaho A Endo, Kyosuke Uno, Hiroyuki Abe, Ryohei Yasuda
ABSTRACT
Oviduct fluid is essential for the fertilization and subsequent preimplantation development. Glycine is abundant in oviduct fluid and is reported to be critical for preimplantation development of fertilized eggs in mammals. However, the mechanism by which glycine exerts its action on fertilized eggs is yet to be understood. Here we show that glycine regulates the preimplantation development of mouse fertilized eggs via glycine receptors. Among them, the alpha-4 subunit (Glra4) and the β subunit are expressed in mouse fertilized eggs, and lacking Glra4 inhibits embryonic development to the blastocyst stage, decreases the number of cells in the blastocysts and the litter size. Thus, we identify a novel function of the glycine receptor, which is considered to act mainly as a neurotransmitter receptor, as a regulator of embryonic development and our data provide new insights into the interactions between oviduct milieu and mammalian fertilized egg.
Sigma-Aldrich
Anti-GLRA4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution