Merck
  • Caspase-4 directly activates caspase-9 in endoplasmic reticulum stress-induced apoptosis in SH-SY5Y cells.
Caspase-4 directly activates caspase-9 in endoplasmic reticulum stress-induced apoptosis in SH-SY5Y cells.
Journal of pharmacological sciences (2011-02-02)
Akiko Yamamuro, Takashi Kishino, Yu Ohshima, Yasuhiro Yoshioka, Tomoki Kimura, Atsushi Kasai, Sadaaki Maeda
ABSTRACT
The present study investigated the function of caspase-4 in endoplasmic reticulum (ER) stress-induced apoptosis in human neuronal cell line SH-SY5Y. Tunicamycin, which is known to induce ER stress, activated both caspase-9 and caspase-4, and the activation of caspase-4 preceded that of caspase-9. The caspase-4 inhibitor LEVD-CHO suppressed both the apoptosis and caspase-9 activation. In addition, human recombinant active caspase-4 cleaved wild type and D330A mutant substituted Asp-330 for alanine of human recombinant procaspase-9, but did not cleave D315A mutant substituted Asp-315 for alanine. These results suggest that caspase-4 directly activates caspase-9 by the processing of procaspase-9 at Asp-315 in ER stress-induced neuronal apoptosis.
Sigma-Aldrich
Caspase 4 Inhibitor (ICH-2), ≥95% (HPLC)
Pricing and availability is not currently available.