Merck
  • Human BMP8A suppresses luteinization of rat granulosa cells via the SMAD1/5/8 pathway.
Human BMP8A suppresses luteinization of rat granulosa cells via the SMAD1/5/8 pathway.
Reproduction (Cambridge, England) (2020-01-16)
Fang-Ju Wu, Ying-Wen Wang, Ching-Wei Luo
ABSTRACT
Bone morphogenetic proteins (BMPs) are known to play an indispensable role in preventing the precocious luteinization of granulosa cells within growing ovarian follicles. In this study, we found that the transcripts of BMP8 genes are enriched in the ovaries of humans and rodents. When analyzing transcriptomic datasets obtained from human mature granulosa cells, we further found that the BMP8 transcripts not only show the highest abundance among the searchable BMP-related ligands but also decrease significantly in women of advanced age or women with polycystic ovarian syndrome. The correlation between the BMP8 levels in granulosa cells and the decline in ovarian function in these subjects suggests that BMP8 protein may be involved in the regulation of granulosa cell function(s). Using a rat model, we demonstrated that human BMP8A protein activates the SMAD1/5/8 and the SMAD2/3 pathways simultaneously in both immature and mature granulosa cells. Furthermore, the expression of potential type I and type II receptors used by BMP8 in rat granulosa cells was characterized. We found that BMP8A treatment can significantly inhibit gonadotropin-induced progesterone production and steroidogenesis-related gene expression in granulosa cells. Pathway dissection using receptor inhibitors further revealed that such inhibitory effects occur specifically through the BMP8-activated SMAD1/5/8, but not SMAD2/3, pathway. Taken together, considering its abundance and possible functions in granulosa cells, we suggest that BMP8 may act as a novel luteinization inhibitor in growing follicles.
Product Description
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Monoclonal Anti-Progesterone antibody produced in rat, clone 2H4, tissue culture supernatant