Merck
  • Frame-shifted amyloid precursor protein found in Alzheimer's disease and Down's syndrome increases levels of secreted amyloid beta40.
Frame-shifted amyloid precursor protein found in Alzheimer's disease and Down's syndrome increases levels of secreted amyloid beta40.
Journal of neurochemistry (2004-07-17)
R van Dijk, D F Fischer, J A Sluijs, M A F Sonnemans, B Hobo, L Mercken, D M A Mann, E M Hol, F W van Leeuwen
ABSTRACT
Frame-shifted amyloid precursor protein (APP(+1)), which has a truncated out-of-frame C-terminus, accumulates in the neuropathological hallmarks of patients with Alzheimer's disease pathology. To study a possible involvement of APP(+1) in the pathogenesis of Alzheimer's disease, we expressed APP695 and APP(+1) in the HEK293 cell-line and studied whether the processing of APP695 was affected. APP(+1) is a secretory protein, but high expression of APP695 and APP(+1) results in the formation of intracellular aggregate-like structures containing both proteins and Fe65, an adaptor protein that interacts with APP695. APP(+1) is shown to interact with APP695, suggesting that these structures consist of functional protein complexes. Such an interaction can also be anticipated in post-mortem brains of young Down's syndrome patients without any sign of neuropathology. Here we observed APP(+1) immunoreactivity in beaded fibres. Additional support for functional consequences on the processing of APP695 comes from a 1.4-fold increase in levels of secreted amyloid beta40 in cells co-expressing APP695 and APP(+1), although APP(+1) itself does not contain the amyloid beta sequence. Taken together, these data show that co-expression of APP695 and APP(+1) affects the processing of APP695 in a pro-amyloidogenic way and this could gradually contribute to Alzheimer's disease pathology, as has been implicated in Down's syndrome patients.
Sigma-Aldrich
Anti-Amyloid Precursor Protein Antibody, CT, serum, Chemicon®