Merck
  • Selective loss of phosphoserine aminotransferase 1 (PSAT1) suppresses migration, invasion, and experimental metastasis in triple negative breast cancer.
Selective loss of phosphoserine aminotransferase 1 (PSAT1) suppresses migration, invasion, and experimental metastasis in triple negative breast cancer.
Clinical & experimental metastasis (2019-10-21)
Stephanie Metcalf, Susan Dougherty, Traci Kruer, Nazarul Hasan, Rumeysa Biyik-Sit, Lindsey Reynolds, Brian F Clem
ABSTRACT
Breast cancer is the second leading cause of cancer-related deaths among women and 90% of these mortalities can be attributed to progression to metastatic disease. In particular, triple negative breast cancer (TNBC) is extremely aggressive and frequently metastasizes to multiple organs. As TNBCs are categorized by their lack of hormone receptors, these tumors are very heterogeneous and are immune to most targeted therapies. Metabolic changes are observed in the majority of TNBC and a large proportion upregulate enzymes within the serine synthesis pathway, including phosphoserine aminotransferase 1 (PSAT1). In this report, we investigate the role of PSAT1 in migration and invasion potential in a subset of TNBC cell types. We found that the expression of PSAT1 increases with TNBC clinical grade. We also demonstrate that suppression of PSAT1 or phosphoglycerate dehydrogenase (PHGDH) does not negatively impact cell proliferation in TNBC cells that are not dependent on de novo serine synthesis. However, we observed that suppression of PSAT1 specifically alters the F-actin cytoskeletal arrangement and morphology in these TNBC cell lines. In addition, suppression of PSAT1 inhibits motility and migration in these TNBC cell lines, which is not recapitulated upon loss of PHGDH. PSAT1 silencing also reduced the number of lung tumor nodules in a model of experimental metastasis; yet did not decrease anchorage-independent growth. Together, these results suggest that PSAT1 functions to drive migratory potential in promoting metastasis in select TNBC cells independent of its role in serine synthesis.
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
MISSION® esiRNA, targeting human PHGDH
Sigma-Aldrich
MISSION® esiRNA, targeting human PSAT1