Merck
  • The pivotal role of glycogen synthase kinase 3 (GSK-3) in vomiting evoked by specific emetogens in the least shrew (Cryptotis parva).
The pivotal role of glycogen synthase kinase 3 (GSK-3) in vomiting evoked by specific emetogens in the least shrew (Cryptotis parva).
Neurochemistry international (2019-11-19)
W Zhong, N A Darmani
ABSTRACT
Glycogen synthase kinase 3 (GSK-3) is a constitutively active multifunctional serine-threonine kinase which is involved in diverse physiological processes. GSK-3 has been implicated in a wide range of diseases including neurodegeneration, inflammation, diabetes and cancer. GSK-3 is a downstream target for protein kinase B (Akt) which phosphorylates GSK-3 and suppresses its activity. Based upon our preliminary findings, we postulated Akt's involvement in emesis. The aim of this study was to investigate the participation of GSK-3 and the antiemetic potential of two GSK-3 inhibitors (AR-A014418 and SB216763) in the least shrew model of vomiting against fully-effective emetic doses of diverse emetogens, including the nonselective and/or selective agonists of serotonin type 3 (e.g. 5-HT or 2-Methyl-5-HT)-, neurokinin type 1 receptor (e.g. GR73632), dopamine D2 (e.g. apomorphine or quinpirole)-, and muscarinic 1 (e.g. pilocarpine or McN-A-343) receptors, as well as the L-type Ca2+ channel agonist (FPL64176), the sarco/endoplasmic reticulum Ca2+-ATPase inhibitor thapsigargin, and the chemotherapeutic agent, cisplatin. We first determined if these emetogens could regulate the phosphorylation level of GSK-3 in the brainstem emetic loci of least shrews and then investigated whether AR-A014418 and SB216763 could protect against the evoked emesis. Phospho-GSK-3α/β Ser21/9 levels in the brainstem and the enteric nerves of jejunum in the small intestine were upregulated following intraperitoneal (i.p.) administration of all the tested emetogens. Furthermore, administration of AR-A014418 (2.5-20 mg/kg, i.p.) dose-dependently attenuated both the frequency and percentage of shrews vomiting in response to i.p. administration of 5-HT (5 mg/kg), 2-Methyl-5-HT (5 mg/kg), GR73632 (5 mg/kg), apomorphine (2 mg/kg), quinpirole (2 mg/kg), pilocarpine (2 mg/kg), McN-A-343 (2 mg/kg), FPL64176 (10 mg/kg), or thapsigargin (0.5 mg/kg). Relatively lower doses of SB216763 exerted antiemetic efficacy, but both inhibitors barely affected cisplatin (10 mg/kg)-induced vomiting. Collectively, these results support the notion that vomiting is accompanied by a downregulation of GSK-3 activity and pharmacological inhibition of GSK-3 protects against pharmacologically evoked vomiting.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film