Merck
  • Astragaloside IV protects ATDC5 cells from lipopolysaccharide-caused damage through regulating miR-203/MyD88.
Astragaloside IV protects ATDC5 cells from lipopolysaccharide-caused damage through regulating miR-203/MyD88.
Pharmaceutical biology (2020-01-08)
Dexin Li, Guangcheng Li, Yang Chen, Yifei Li, Junfeng Zhang, Dexuan Gao, Linglong Sun, Bo Liu
ABSTRACT
Context: Osteoarthritis (OA) is a degenerative arthrosis sickness. Astragaloside IV (AS-IV) functions by relieving inflammatory damage.Objective: We aimed to investigate the mechanism by which AS-IV protects ATD cells from lipopolysaccharide (LPS)-induced damage.Materials and methods: ATDC5 cells were transfected with miR-203 inhibitor and NC inhibitor (150 nM) or pEX-MyD88 and sh-MyD88 (50 nM) for 48 h, pre-treated by 15 μg/mL AS-IV for 24 h, then treated by 5 μg/mL LPS for 12 h. Dual-luciferase activity testing was used to determine whether miR-203 could bind to MyD88. CCK-8 and flow cytometry were used to detect cell activity and apoptosis, respectively, and qRT-PCR, western blots, and ELISA were performed to detect expression levels of miR-203 and inflammatory cytokines.Results: Based on the 50% inhibiting concentration (IC50), there was no significant difference of AS-IV (0 to 15 μg/mL) on cell viability. Fifteen μg/mL was the optimal concentration of AS-IV in treating LPS-induced inflammatory damage in subsequent experiments since this was a semi-lethal concentration. AS-IV significantly reduces LPS-induced viability, apoptosis and the release of TNF-α, IL-6 and iNOS mainly through up-regulating miR-203. Further, MyD88 was a target gene of miR-203 and negatively regulated by miR-203. Knockdown of MyD88 inhibited LPS-induced inflammatory damage by inhibiting the NF-κB signal pathway.Discussion and conclusions: AS-IV protects ATDC5 cells against LPS-induced damage mainly via regulating miR-203/MyD88. Our results support a theoretical basis for in-depth study of the function of AS-IV and the clinical cure of OA.
Product Description
Millipore
Astragaloside IV, >98.0%
Pricing and availability is not currently available.