Merck
  • CHEK1 coordinates DNA damage signaling and meiotic progression in the male germline of mice.
CHEK1 coordinates DNA damage signaling and meiotic progression in the male germline of mice.
Human molecular genetics (2018-01-24)
Hironori Abe, Kris G Alavattam, Yasuko Kato, Diego H Castrillon, Qishen Pang, Paul R Andreassen, Satoshi H Namekawa
ABSTRACT
The continuity of life depends on mechanisms in the germline that ensure the integrity of the genome. The DNA damage response/checkpoint kinases ATM and ATR are essential signaling factors in the germline. However, it remains unknown how a downstream transducer, Checkpoint Kinase 1 (CHEK1 or CHK1), mediates signaling in the male germline. Here, we show that CHEK1 has distinct functions in both the mitotic and meiotic phases of the male germline in mice. In the mitotic phase, CHEK1 is required for the resumption of prospermatogonia proliferation after birth and the maintenance of spermatogonia. In the meiotic phase, we uncovered two functions for CHEK1: one is the stage-specific attenuation of DNA damage signaling on autosomes, and the other is coordination of meiotic stage progression. On autosomes, the loss of CHEK1 delays the removal of DNA damage signaling that manifests as phosphorylation of histone variant H2AX at serine 139 (γH2AX). Importantly, CHEK1 does not have a direct function in meiotic sex chromosome inactivation (MSCI), an essential event in male meiosis, in which ATR is a key regulator. Thus, the functions of ATR and CHEK1 are uncoupled in MSCI, in contrast to their roles in DNA damage signaling in somatic cells. Our study reveals stage-specific functions for CHEK1 that ensure the integrity of the male germline.
Sigma-Aldrich
Anti-Chk1 antibody, Mouse monoclonal, ~2 mg/mL, clone DCS-310, purified from hybridoma cell culture