Merck
  • Palmitate and oleate exert differential effects on insulin signalling and glucose uptake in human skeletal muscle cells.
Palmitate and oleate exert differential effects on insulin signalling and glucose uptake in human skeletal muscle cells.
Endocrine connections (2017-06-07)
Selina Mäkinen, Yen H Nguyen, Paulina Skrobuk, Heikki A Koistinen
ABSTRACT
Saturated fatty acids are implicated in the development of insulin resistance, whereas unsaturated fatty acids may have a protective effect on metabolism. We tested in primary human myotubes if insulin resistance induced by saturated fatty acid palmitate can be ameliorated by concomitant exposure to unsaturated fatty acid oleate. Primary human myotubes were pretreated with palmitate, oleate or their combination for 12 h. Glucose uptake was determined by intracellular accumulation of [3H]-2-deoxy-d-glucose, insulin signalling and activation of endoplasmic reticulum (ER) stress by Western blotting, and mitochondrial reactive oxygen species (ROS) production by fluorescent dye MitoSOX. Exposure of primary human myotubes to palmitate impaired insulin-stimulated Akt-Ser473, AS160 and GSK-3β phosphorylation, induced ER stress signalling target PERK and stress kinase JNK 54 kDa isoform. These effects were virtually abolished by concomitant exposure of palmitate-treated myotubes to oleate. However, an exposure to palmitate, oleate or their combination reduced insulin-stimulated glucose uptake. This was associated with increased mitochondrial ROS production in palmitate-treated myotubes co-incubated with oleate, and was alleviated by antioxidants MitoTempo and Tempol. Thus, metabolic and intracellular signalling events diverge in myotubes treated with palmitate and oleate. Exposure of human myotubes to excess fatty acids increases ROS production and induces insulin resistance.
Sigma-Aldrich
Anti-phospho-AMPKα (Thr172) Antibody, Upstate®, from rabbit