Merck
  • Moxibustion at CV4 alleviates atherosclerotic lesions through activation of the LXRα/ABCA1 pathway in apolipoprotein-E-deficient mice.
Moxibustion at CV4 alleviates atherosclerotic lesions through activation of the LXRα/ABCA1 pathway in apolipoprotein-E-deficient mice.
Acupuncture in medicine : journal of the British Medical Acupuncture Society (2019-05-30)
Yingxue Cui, Juntian Liu, Chang Huang, Baixiao Zhao
ABSTRACT
To investigate the anti-atherogenic effect of moxibustion and whether it is mediated through the reverse cholesterol transport process. 8-week-old male apolipoprotein E deficient (ApoE-/- knockout) mice were randomly divided into two groups (n=10 per group): atherosclerosis (AS) and AS plus moxibustion (AS+M). C57BL/6J mice of the same background (n=10) were selected as controls. Mice in the AS+M group received indirect moxibustion with an ignited moxa stick held over CV4. Mice of the AS and control groups were restrained in the same holder with an unlit moxa stick held over CV4. All treatments were performed for 20 min per day, 6 days per week for 12 weeks. After the treatment, the mice were euthanased and their serum lipids were measured. The aortic roots and thoracic aortas were collected for haematoxylin and eosin and red oil O staining, respectively, to analyse the atherosclerotic lesions. Expression of adenosine triphosphate binding cassette (ABCA)A1/G1 and liver X receptor α (LXRα) in the thoracic aorta were examined with Western blotting. The moxibustion-treated (AS+M) mice showed a significantly lower plaque area percentage in the aortic root and thoracic aorta, and higher expression of LXRα and ABCA1 in the thoracic aorta compared with the AS mice. No significant differences were found in average lipid area percentage in the thoracic aorta, or ABCG1 expression in the thoracic aorta, between mice in the AS+M and AS groups. Moxibustion treatment at CV4 suppressed the progression of atherosclerotic lesions in ApoE-/- mice. The anti-atherogenic effect of moxibustion may be achieved by: (1) regulation of lipid metabolism, and thus prevention of lipid accumulation; and (2) upregulation of LXRα- and ABCA1-mediated cholesterol efflux in the lesion area.