Merck
  • Bacterial biofilm in adenoids of children with chronic otitis media. Part I: a case control study of prevalence of biofilms in adenoids, risk factors and middle ear biofilms.
Bacterial biofilm in adenoids of children with chronic otitis media. Part I: a case control study of prevalence of biofilms in adenoids, risk factors and middle ear biofilms.
Acta oto-laryngologica (2019-02-27)
Romain Kania, Pierre Vironneau, Huong Dang, Béatrice Bercot, Emmanuelle Cambau, Benjamin Verillaud, Domitille Camous, Gerda Lamers, Philippe Herman, Eric Vicaut, Natacha Tessier, Thierry Van Den Abbeele
ABSTRACT
Biofilms are communities of bacteria embedded in a self-produced glycocalyx matrix. Adenoids have been shown to harbor bacterial biofilms. Aim/objectives: To compare the prevalence of biofilms in adenoid of children with chronic otitis media (COM) (group1) versus a control group without any COM (group 2) having adenoids removed because of hypertrophy. One hundred and three children were prospectively enrolled in this case-control study, group 1 (n = 52) and group 2 (n = 51). The main outcome measurement was the prevalence of biofilm in adenoidectomy specimens analyzed using confocal laser scanning microscopy. Children in group 1 who had middle ear (ME) effusion and requiring the insertion of a tympanostomy tube underwent biopsy of the ME mucosa and effusion sampling. Biofilms were found in adenoids' specimens of both groups and in the ME biopsy and effusion. The biofilm prevalence in adenoids was 63.5% (33/52) in group 1 and 47.1% (24/51) in group 2. Day nursery and previous antibiotics intake were significantly more frequent in group 1 than in group 2. This case-control study demonstrates that adenoid tissue in children with COM contains more mucosal biofilms than adenoid tissue removed for hypertrophy. Biofilm was seen in ME biopsies and effusion.