Merck
  • Myosin 1F Regulates M1-Polarization by Stimulating Intercellular Adhesion in Macrophages.
Myosin 1F Regulates M1-Polarization by Stimulating Intercellular Adhesion in Macrophages.
Frontiers in immunology (2019-01-29)
Zayda L Piedra-Quintero, Carolina Serrano, Nicolás Villegas-Sepúlveda, José L Maravillas-Montero, Sandra Romero-Ramírez, Mineko Shibayama, Oscar Medina-Contreras, Porfirio Nava, Leopoldo Santos-Argumedo
ABSTRACT
Intestinal macrophages are highly mobile cells with extraordinary plasticity and actively contribute to cytokine-mediated epithelial cell damage. The mechanisms triggering macrophage polarization into a proinflammatory phenotype are unknown. Here, we report that during inflammation macrophages enhance its intercellular adhesion properties in order to acquire a M1-phenotype. Using in vitro and in vivo models we demonstrate that intercellular adhesion is mediated by integrin-αVβ3 and relies in the presence of the unconventional class I myosin 1F (Myo1F). Intercellular adhesion mediated by αVβ3 stimulates M1-like phenotype in macrophages through hyperactivation of STAT1 and STAT3 downstream of ILK/Akt/mTOR signaling. Inhibition of integrin-αVβ3, Akt/mTOR, or lack of Myo1F attenuated the commitment of macrophages into a pro-inflammatory phenotype. In a model of colitis, Myo1F deficiency strongly reduces the secretion of proinflammatory cytokines, decreases epithelial damage, ameliorates disease activity, and enhances tissue repair. Together our findings uncover an unknown role for Myo1F as part of the machinery that regulates intercellular adhesion and polarization in macrophages.
Sigma-Aldrich
Anti-MYO1F antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Cilengitide trifluoroacetic acid salt, ≥95% (HPLC)
Pricing and availability is not currently available.