Merck
  • CDK1-CCNB1 creates a spindle checkpoint-permissive state by enabling MPS1 kinetochore localization.
CDK1-CCNB1 creates a spindle checkpoint-permissive state by enabling MPS1 kinetochore localization.
The Journal of cell biology (2019-01-25)
Daniel Hayward, Tatiana Alfonso-Pérez, Michael J Cundell, Michael Hopkins, James Holder, James Bancroft, Lukas H Hutter, Bela Novak, Francis A Barr, Ulrike Gruneberg
ABSTRACT
Spindle checkpoint signaling is initiated by recruitment of the kinase MPS1 to unattached kinetochores during mitosis. We show that CDK1-CCNB1 and a counteracting phosphatase PP2A-B55 regulate the engagement of human MPS1 with unattached kinetochores by controlling the phosphorylation status of S281 in the kinetochore-binding domain. This regulation is essential for checkpoint signaling, since MPS1S281A is not recruited to unattached kinetochores and fails to support the recruitment of other checkpoint proteins. Directly tethering MPS1S281A to the kinetochore protein Mis12 bypasses this regulation and hence the requirement for S281 phosphorylation in checkpoint signaling. At the metaphase-anaphase transition, MPS1 S281 dephosphorylation is delayed because PP2A-B55 is negatively regulated by CDK1-CCNB1 and only becomes fully active once CCNB1 concentration falls below a characteristic threshold. This mechanism prolongs the checkpoint-responsive period when MPS1 can localize to kinetochores and enables a response to late-stage spindle defects. By acting together, CDK1-CCNB1 and PP2A-B55 thus create a spindle checkpoint-permissive state and ensure the fidelity of mitosis.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Cyclin B1 Antibody, clone GNS3 (8A5D12), clone GNS3 (8A5D12), Upstate®, from mouse