Merck
Cell entry of a host-targeting protein of oomycetes requires gp96.
Nature communications (2018-06-16)
Franziska Trusch, Lars Loebach, Stephan Wawra, Elaine Durward, Andreas Wuensch, Nurul Aqilah Iberahim, Irene de Bruijn, Kevin MacKenzie, Ariane Willems, Aleksandra Toloczko, Javier Diéguez-Uribeondo, Tim Rasmussen, Thomas Schrader, Peter Bayer, Chris J Secombes, Pieter van West
ABSTRACT
The animal-pathogenic oomycete Saprolegnia parasitica causes serious losses in aquaculture by infecting and killing freshwater fish. Like plant-pathogenic oomycetes, S. parasitica employs similar infection structures and secretes effector proteins that translocate into host cells to manipulate the host. Here, we show that the host-targeting protein SpHtp3 enters fish cells in a pathogen-independent manner. This uptake process is guided by a gp96-like receptor and can be inhibited by supramolecular tweezers. The C-terminus of SpHtp3 (containing the amino acid sequence YKARK), and not the N-terminal RxLR motif, is responsible for the uptake into host cells. Following translocation, SpHtp3 is released from vesicles into the cytoplasm by another host-targeting protein where it degrades nucleic acids. The effector translocation mechanism described here, is potentially also relevant for other pathogen-host interactions as gp96 is found in both animals and plants.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Pricing and availability is not currently available.